MilliporeSigma
  • Prohibition or coffee shops: regulation of amphetamine and methylphenidate for enhancement use by healthy adults.
Prohibition or coffee shops: regulation of amphetamine and methylphenidate for enhancement use by healthy adults.
The American journal of bioethics : AJOB (2013-06-19)
Veljko Dubljević
ABSTRACT
This article analyzes appropriate public policies for enhancement use of two most important stimulant drugs: Ritalin (methylphenidate) and Adderall (mixed amphetamine salts). The author argues that appropriate regulation of cognition enhancement drugs cannot be a result of a general discussion on cognitive enhancements as such, but has to be made on a case-by-case basis. Starting from the recently proposed taxation approach to cognition enhancement drugs, the author analyzes available, moderately permissive models of regulation. After a thorough analysis of relevant characteristics of methylphenidate and amphetamine, the author concludes that a moderately liberal permissive regulation of enhancement use by healthy adults might be appropriate for extended release forms of methylphenidate. However, due to their danger profile, amphetamine and instant release forms of methylphenidate should not be made readily available to healthy adults and would need to be prohibited.
Product Description
Supelco
(±)-Amphetamine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®
Supelco
Methylphenidate hydrochloride solution, (Racemic mixture), ampule of 1 mL, 1.0 mg/mL in methanol (as free base), certified reference material, Cerilliant®