MilliporeSigma
  • RNAtips: Analysis of temperature-induced changes of RNA secondary structure.
RNAtips: Analysis of temperature-induced changes of RNA secondary structure.
Nucleic acids research (2013-06-15)
Andrey Chursov, Sebastian J Kopetzky, Gennady Bocharov, Dmitrij Frishman, Alexander Shneider
ABSTRACT
Although multiple biological phenomena are related to temperature (e.g. elevation of body temperature due to an illness, adaptation to environmental temperature conditions, biology of coldblooded versus warm-blooded organisms), the molecular mechanisms of these processes remain to be understood. Perturbations of secondary RNA structures may play an important role in an organism's reaction to temperature change--in all organisms from viruses and bacteria to humans. Here, we present RNAtips (temperature-induced perturbation of structure) web server, which can be used to predict regions of RNA secondary structures that are likely to undergo structural alterations prompted by temperature change. The server can also be used to: (i) detect those regions in two homologous RNA sequences that undergo different structural perturbations due to temperature change and (ii) test whether these differences are specific to the particular nucleotide substitutions distinguishing the sequences. The RNAtips web server is freely accessible without any login requirement at http://rnatips.org.
Sigma-Aldrich
Ribonucleic acid diethylaminoethanol salt, Type IX
Sigma-Aldrich
Ribonucleic acid from baker′s yeast (S. cerevisiae)
Sigma-Aldrich
Ribonucleic acid from torula yeast, Type VI
Sigma-Aldrich
Ribonucleic acid from torula yeast, core, Type II-C