MilliporeSigma
  • Transient proteolytic modification of mesenchymal stromal cells increases lung clearance rate and targeting to injured tissue.
Transient proteolytic modification of mesenchymal stromal cells increases lung clearance rate and targeting to injured tissue.
Stem cells translational medicine (2013-06-05)
Erja Kerkelä, Tanja Hakkarainen, Tuomas Mäkelä, Mari Raki, Oleg Kambur, Lotta Kilpinen, Janne Nikkilä, Siri Lehtonen, Ilja Ritamo, Roni Pernu, Mika Pietilä, Reijo Takalo, Tatu Juvonen, Kim Bergström, Eija Kalso, Leena Valmu, Saara Laitinen, Petri Lehenkari, Johanna Nystedt
ABSTRACT
Systemic infusion of therapeutic cells would be the most practical and least invasive method of administration in many cellular therapies. One of the main obstacles especially in intravenous delivery of cells is a massive cell retention in the lungs, which impairs homing to the target tissue and may decrease the therapeutic outcome. In this study we showed that an alternative cell detachment of mesenchymal stromal/stem cells (MSCs) with pronase instead of trypsin significantly accelerated the lung clearance of the cells and, importantly, increased their targeting to an area of injury. Cell detachment with pronase transiently altered the MSC surface protein profile without compromising cell viability, multipotent cell characteristics, or immunomodulative and angiogenic potential. The transient modification of the cell surface protein profile was sufficient to produce effective changes in cell rolling behavior in vitro and, importantly, in the in vivo biodistribution of the cells in mouse, rat, and porcine models. In conclusion, pronase detachment could be used as a method to improve the MSC lung clearance and targeting in vivo. This may have a major impact on the bioavailability of MSCs in future therapeutic regimes.
Sigma-Aldrich
Carrageenan, suitable for gel preparation