Merck
  • [Study on effective constituents extracted from fibrous roots of Salvia miltiorrhiza with degrading multi-enzymes from taishan Ganoderma lucidum].
[Study on effective constituents extracted from fibrous roots of Salvia miltiorrhiza with degrading multi-enzymes from taishan Ganoderma lucidum].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-06-04)
Yan-Ling Li, Xiao-Ming Xin, Zeng-Min Miao, Ren-Jiu Shi, Gang-Ping Hao
ABSTRACT
To study the application of degrading multi-enzymes from Ganoderma lucidum in extracting effective constituents from fibrous roots of Salvia miltiorrhiza. Effective constituents were extracted from fibrous roots by degrading multi-enzymes of wood fiber. The enzymatic parameters were optimized by the orthogonal design. The extraction efficiencies of total tanshinones and total salvianolic acids in the extracts of fibrous roots of S. miltiorrhiza was obtained using optimum enzymolysis process reached 11.923%, 12.465%, respectively, which were 62.794%, 56.086% more than that by conventional non-enzymatic hydrolysis. Degrading multi-enzymes of wood fiber can be used to fully extract effective constituents from fibrous roots of S. miltiorrhiza, which provides a new approach for recycling wastes of traditional Chinese medicines.
Sigma-Aldrich
Tanshinone I, ≥98% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$491.00
Sigma-Aldrich
Tanshinone IIA, ≥97% (HPLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
10 mg
In Stock
Details...
$491.00