MilliporeSigma
  • Candida tropicalis: diagnostic dilemmas for an unusual prosthetic hip infection.
Candida tropicalis: diagnostic dilemmas for an unusual prosthetic hip infection.
Journal of the Royal Army Medical Corps (2013-05-31)
Surjit Lidder, A Tasleem, S Masterson, R W J Carrington
ABSTRACT
Infection is the third commonest cause of total hip arthroplasty failure. Infections of the hip with Candida species are extremely rare with only a few reports in the literature. A case of a 76-year-old female subject is presented illustrating both the difficulty in initial diagnosis and the challenges faced in hip reconstruction.
Sigma-Aldrich
Amphotericin B solubilized, powder, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., BioReagent, suitable for cell culture, ~80% (HPLC)
Sigma-Aldrich
Amphotericin B solution, 250 μg/mL in deionized water, 0.1 μm filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Amphotericin B from Streptomyces sp., ~80% (HPLC), powder
Amphotericin B for peak identification, European Pharmacopoeia (EP) Reference Standard