MilliporeSigma
The absorption of iron from whole diets: a systematic review.
The American journal of clinical nutrition (2013-05-31)
Rachel Collings, Linda J Harvey, Lee Hooper, Rachel Hurst, Tracey J Brown, Jennifer Ansett, Maria King, Susan J Fairweather-Tait
ABSTRACT
Absorption factors are required to convert physiologic requirements for iron into Dietary Reference Values, but the absorption from single meals cannot be used to estimate dietary iron absorption. The objective was to conduct a systematic review of iron absorption from whole diets. A structured search was completed by using the Medline, EMBASE, and Cochrane CENTRAL databases from inception to November 2011. Formal inclusion and exclusion criteria were applied, and data extraction, validity assessment, and meta-analyses were undertaken. Nineteen studies from the United States, Europe, and Mexico were included. Absorption from diets was higher with an enhancer (standard mean difference: 0.53; 95% CI: 0.21, 0.85; P = 0.001) and was also higher when compared with low-bioavailability diets (standard mean difference: 0.96; 95% CI: 0.51, 1.41; P < 0.0001); however, single inhibitors did not reduce absorption (possibly because of the limited number of studies and participants and their heterogeneity). A regression equation to calculate iron absorption was derived by pooling data for iron status (serum and plasma ferritin) and dietary enhancers and inhibitors from 58 individuals (all from US studies): log[nonheme-iron absorption, %] = -0.73 log[ferritin, μg/L] + 0.11 [modifier] + 1.82. In individuals with serum ferritin concentrations from 6 to 80 μg/L, predicted absorption ranged from 2.1% to 23.0%. Large variations were observed in mean nonheme-iron absorption (0.7-22.9%) between studies, which depended on iron status (diet had a greater effect at low serum and plasma ferritin concentrations) and dietary enhancers and inhibitors. Iron absorption was predicted from serum ferritin concentrations and dietary modifiers by using a regression equation. Extrapolation of these findings to developing countries and to men and women of different ages will require additional high-quality controlled trials.
Sigma-Aldrich
Ferritin from human spleen, Type V, 10 μg/mL in 0.15 M NaCl, 10 mM Tris, pH 8.0, containing 0.1% sodium azide
Sigma-Aldrich
Ferritin from equine spleen, Type I, saline solution
Sigma-Aldrich
Ferritin from human liver, Type IV, 10 μg/mL