MilliporeSigma
  • Dynamic methylation of Numb by Set8 regulates its binding to p53 and apoptosis.
Dynamic methylation of Numb by Set8 regulates its binding to p53 and apoptosis.
Molecular cell (2013-05-28)
Gurpreet Kaur Dhami, Huadong Liu, Marek Galka, Courtney Voss, Ran Wei, Kimberly Muranko, Tomonori Kaneko, Sean P Cregan, Lin Li, Shawn Shun-Cheng Li
ABSTRACT
Although Numb exhibits its tumor-suppressive function in breast cancer in part by binding to and stabilizing p53, it is unknown how the Numb-p53 interaction is regulated in cells. We found that Numb is methylated in its phosphotyrosine-binding (PTB) domain by the lysine methyltransferase Set8. Moreover, methylation uncouples Numb from p53, resulting in increased p53 ubiquitination and degradation. While Numb promotes apoptosis in a p53-dependent manner, the apoptotic function is abolished when Numb is methylated by Set8 or the Lys methylation sites in Numb are mutated. Conversely, the Numb-p53 interaction and Numb-mediated apoptosis are significantly enhanced by depletion of Set8 from cancer cells or by treating the cells with doxorubicin, a chemotherapeutic drug that causes a reduction in the mRNA and protein levels of Set8. Our work identifies the Set8-Numb-p53 signaling axis as an important regulatory pathway for apoptosis and suggests a therapeutic strategy by targeting Numb methylation.
Sigma-Aldrich
L-Lysine, ≥98% (TLC)
Sigma-Aldrich
L-Lysine acetate salt, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine, crystallized, ≥98.0% (NT)
Supelco
L-Lysine monohydrochloride, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
L-Lysine monohydrochloride, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Supelco
L-Lysine Acetate, Pharmaceutical Secondary Standard; Certified Reference Material