MilliporeSigma
  • Glutathione and tryptophan metabolism are required for Arabidopsis immunity during the hypersensitive response to hemibiotrophs.
Glutathione and tryptophan metabolism are required for Arabidopsis immunity during the hypersensitive response to hemibiotrophs.
Proceedings of the National Academy of Sciences of the United States of America (2013-05-23)
Kei Hiruma, Satoshi Fukunaga, Pawel Bednarek, Mariola Pislewska-Bednarek, Satoshi Watanabe, Yoshihiro Narusaka, Ken Shirasu, Yoshitaka Takano
ABSTRACT
The hypersensitive response (HR) is a type of strong immune response found in plants that is accompanied by localized cell death. However, it is unclear how HR can block a broad range of pathogens with different infective modes. In this study, we report that γ-glutamylcysteine synthetase GSH1, which is critical for glutathione biosynthesis, and tryptophan (Trp) metabolism contribute to HR and block development of fungal pathogens with hemibiotrophic infective modes. We found that GSH1 is involved in the penetration2 (PEN2)-based entry control of the nonadapted hemibiotroph Colletotrichum gloeosporioides. However, Arabidopsis mutants specifically defective in entry control terminated further growth of the pathogen in the presence of HR cell death, whereas gsh1 mutants supported pathogen invasive growth in planta, demonstrating the requirement of GSH1 for postinvasive nonhost resistance. Remarkably, on the basis of the phenotypic and metabolic analysis of Arabidopsis mutants defective in Trp metabolism, we showed that biosynthesis of Trp-derived phytochemicals is also essential for resistance to C. gloeosporioides during postinvasive HR. By contrast, GSH1 and these metabolites are likely to be dispensable for the induction of cell death during postinvasive HR. Furthermore, the resistance to Ralstonia solanacearum 1/resistance to Pseudomonas syringae 4 dual Resistance gene-dependent immunity of Arabidopsis to the adapted hemibiotroph shared GSH1 and cytochromes P450 CYP79B2/CYP79B3 with postinvasive nonhost resistance, whereas resistance to P. syringae pv. maculicola 1 and resistance to P. syringae 2-based Resistance gene resistance against bacterial pathogens did not. These data suggest that the synthesis of glutathione and Trp-derived metabolites during HR play crucial roles in terminating the invasive growth of both nonadapted and adapted hemibiotrophs.
Product Description
Supelco
Tryptophan, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland