MilliporeSigma
  • The antiprion compound 6-aminophenanthridine inhibits the protein folding activity of the ribosome by direct competition.
The antiprion compound 6-aminophenanthridine inhibits the protein folding activity of the ribosome by direct competition.
The Journal of biological chemistry (2013-05-16)
Yanhong Pang, Sriram Kurella, Cécile Voisset, Dibyendu Samanta, Debapriya Banerjee, Ariane Schabe, Chanchal Das Gupta, Hervé Galons, Marc Blondel, Suparna Sanyal
ABSTRACT
Domain V of the 23S/25S/28S rRNA of the large ribosomal subunit constitutes the active center for the protein folding activity of the ribosome (PFAR). Using in vitro transcribed domain V rRNAs from Escherichia coli and Saccharomyces cerevisiae as the folding modulators and human carbonic anhydrase as a model protein, we demonstrate that PFAR is conserved from prokaryotes to eukaryotes. It was shown previously that 6-aminophenanthridine (6AP), an antiprion compound, inhibits PFAR. Here, using UV cross-linking followed by primer extension, we show that the protein substrates and 6AP interact with a common set of nucleotides on domain V of 23S rRNA. Mutations at the interaction sites decreased PFAR and resulted in loss or change of the binding pattern for both the protein substrates and 6AP. Moreover, kinetic analysis of human carbonic anhydrase refolding showed that 6AP decreased the yield of the refolded protein but did not affect the rate of refolding. Thus, we conclude that 6AP competitively occludes the protein substrates from binding to rRNA and thereby inhibits PFAR. Finally, we propose a scheme clarifying the mechanism by which 6AP inhibits PFAR.
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, Isoelectric focusing marker, pI 5.4
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II from bovine erythrocytes, lyophilized powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase Isozyme II human, ≥80%, powder, ≥3,000 W-A units/mg protein
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, ≥95% (SDS-PAGE), specific activity ≥3,500 W-A units/mg protein, lyophilized powder
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, For use as a marker in SDS-PAGE
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, non-denaturing PAGE marker
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, lyophilized powder, ≥2,000 W-A units/mg protein
Supelco
Carbonic Anhydrase I from human erythrocytes, Isoelectric focusing marker, pI 6.6
Sigma-Aldrich
Carbonic Anhydrase I from human erythrocytes
Sigma-Aldrich
Carbonic Anhydrase from bovine erythrocytes, BioReagent, suitable for GFC marker