MilliporeSigma
[Chemical constituents of Gentiana rhodantha].
Zhongguo Zhong yao za zhi = Zhongguo zhongyao zazhi = China journal of Chinese materia medica (2013-05-15)
Yun Chen, Guo-Kai Wang, Can Wu, Min-Jian Qin
ABSTRACT
To determine the chemical constituents of Gentiana rhodantha. To isolate the constituents, column chromatography over silica gel, MCI, Sephadex LH-20 and C18 reverse-phased silica gel were used. Spectroscopic methods were used to elucidate the structures of the isolated compounds. Sixteen compounds were isolated and elucidated as ten phonemic compounds, namely 1,3,7,8-tetrahydroxylxanthone (1), rhodanthenone D (2), 1,3,6,7-tetrahydroxylxanthone (3), 1,3,7-trihydroxy-4,8-dimethoxyxanthone (4), quercetin (5), isoorientin (6), mangiferin (7), norswertianolin (8), gallic acid ethyl ester (9) and salicylic acid (10), and six triterpenes including alpha-amyrin (11), erythrodiol 3-O-palmitate (12), ursolic aldehyde (13), uvaol 3-O-acetyl (14), ursolic acid (15) and 2alpha-hydroxyursolic acid (16). Compounds 4-6, 8, 10-12, 15 and 16 were isolated from this plant for the first time. Compounds 1 and 3 were obtained firstly from the genus Gentiana and compounds 9, 13-14 were firstly from the family Gentianaceae.
Product Description
Supelco
Mangiferin, analytical standard
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~100,000
Salicylic acid, European Pharmacopoeia (EP) Reference Standard
Supelco
Salicylic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dextran, analytical standard, for GPC, Set Mp 1,000-400,000
Supelco
Dextran from Leuconostoc mesenteroides, for GPC, analytical standard, Mw 50,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 670,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~200,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 5,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 25,000
Supelco
Dextran from Leuconostoc mesenteroides, analytical standard, for GPC, Mw 410,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr 450,000-650,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, Mr ~60,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~70,000
Supelco
Salicylic acid, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Sephadex® G-25, BioReagent, for molecular biology, DNA grade, Superfine
Sigma-Aldrich
Salicylic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 35,000-45,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 15,000-30,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 150,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 48,000-90,000
Sigma-Aldrich
Mangiferin Mangifera indica
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 9,000-11,000
Sigma-Aldrich
Dextran from Leuconostoc mesenteroides, average mol wt 1,500,000-2,800,000
Sigma-Aldrich
Dextran from Leuconostoc spp., Mr ~6,000
Sigma-Aldrich
Sephadex® G-75, Superfine
Sigma-Aldrich
Sephadex® G-100, Medium
Sigma-Aldrich
Sephadex® G-15, Medium
Sigma-Aldrich
Sephadex® G-25, Medium
Sigma-Aldrich
Sephadex® G-25, Fine