MilliporeSigma
  • Treatment of small cell lung cancer: Diagnosis and management of lung cancer, 3rd ed: American College of Chest Physicians evidence-based clinical practice guidelines.
Treatment of small cell lung cancer: Diagnosis and management of lung cancer, 3rd ed: American College of Chest Physicians evidence-based clinical practice guidelines.
Chest (2013-05-10)
James R Jett, Steven E Schild, Kenneth A Kesler, Gregory P Kalemkerian
ABSTRACT
Small cell lung cancer (SCLC) is a lethal disease for which there have been only small advances in diagnosis and treatment in the past decade. Our goal was to revise the evidence-based guidelines on staging and best available treatment options. A comprehensive literature search covering 2004 to 2011 was conducted in MEDLINE, Embase, and five Cochrane databases using SCLC terms. This was cross-checked with the authors' own literature searches and knowledge of the literature. Results were limited to research in humans and articles written in English. The staging classification should include both the old Veterans Administration staging classification of limited stage (LS) and extensive stage (ES), as well as the new seventh edition American Joint Committee on Cancer/International Union Against Cancer staging by TNM. The use of PET scanning is likely to improve the accuracy of staging. Surgery is indicated for carefully selected stage I SCLC. LS disease should be treated with concurrent chemoradiotherapy in patients with good performance status. Thoracic radiotherapy should be administered early in the course of treatment, preferably beginning with cycle 1 or 2 of chemotherapy. Chemotherapy should consist of four cycles of a platinum agent and etoposide. ES disease should be treated primarily with chemotherapy consisting of a platinum agent plus etoposide or irinotecan. Prophylactic cranial irradiation prolongs survival in those individuals with both LS and ES disease who achieve a complete or partial response to initial therapy. To date, no molecularly targeted therapy agent has demonstrated proven efficacy against SCLC. Evidence-based guidelines are provided for the staging and treatment of SCLC. LS-SCLC is treated with curative intent with 20% to 25% 5-year survival. ES-SCLC is initially responsive to standard treatment, but almost always relapses, with virtually no patients surviving for 5 years. Targeted therapies have no proven efficacy against SCLC.
Sigma-Aldrich
(S)-(+)-Camptothecin, ≥90% (HPLC), powder
Sigma-Aldrich
Irinotecan hydrochloride, topoisomerase inhibitor
Sigma-Aldrich
7-Ethyl-10-hydroxycamptothecin, ≥98% (HPLC), powder
Sigma-Aldrich
Etoposide, synthetic, 95.0-105.0%, powder