MilliporeSigma
  • [Problems of tobacco smoke and positive effects of quit-smoking on human health].
[Problems of tobacco smoke and positive effects of quit-smoking on human health].
Nihon rinsho. Japanese journal of clinical medicine (2013-05-02)
Mayumi Abe
ABSTRACT
Cigarette smoking dominates among Japanese smokers. A smoker takes a small amount of nicotine into the body by one aspiration of cigarette; this amount is not large enough to cause acute symptoms. The way of consuming one cigarette by about 10 aspirations may supply a sufficient amount of nicotine to the body without producing unpleasant symptoms of acute toxicity. Cigarette is very effective goods of nicotine delivery to increase the nicotine-dependent patients. In addition, owing to the tricky image strategy by a tobacco company, the citizens in Japan have not become sufficiently aware of hazards of tobacco. For health hazards of smoking, people do not fully understand cigarette smoking may affect the whole body because of the strong impression of "cancer" particularly "lung cancer" as the disease of smoking. Therefore, we, healthcare professionals, should treat diseases with accurate knowledge of tobacco hazards and also should play a positive role to enlighten people about the truth of tobacco hazards, which do not mean cancer alone.
Product Description
Supelco
(−)-Nicotine solution, 1.0 mg/mL, analytical standard, for drug analysis
Supelco
(−)-Nicotine, PESTANAL®, analytical standard
Sigma-Aldrich
(−)-Nicotine, ≥99% (GC), liquid
Supelco
S(−)-Nicotine solution, 1.0 mg/mL in methanol, ampule of 1 mL, certified reference material, Cerilliant®