MilliporeSigma
  • Role of phosphatidylethanolamine in the biogenesis of mitochondrial outer membrane proteins.
Role of phosphatidylethanolamine in the biogenesis of mitochondrial outer membrane proteins.
The Journal of biological chemistry (2013-04-30)
Thomas Becker, Susanne E Horvath, Lena Böttinger, Natalia Gebert, Günther Daum, Nikolaus Pfanner
ABSTRACT
The mitochondrial outer membrane contains proteinaceous machineries for the import and assembly of proteins, including TOM (translocase of the outer membrane) and SAM (sorting and assembly machinery). It has been shown that the dimeric phospholipid cardiolipin is required for the stability of TOM and SAM complexes and thus for the efficient import and assembly of β-barrel proteins and some α-helical proteins of the outer membrane. Here, we report that mitochondria deficient in phosphatidylethanolamine (PE), the second non-bilayer-forming phospholipid, are impaired in the biogenesis of β-barrel proteins, but not of α-helical outer membrane proteins. The stability of TOM and SAM complexes is not disturbed by the lack of PE. By dissecting the import steps of β-barrel proteins, we show that an early import stage involving translocation through the TOM complex is affected. In PE-depleted mitochondria, the TOM complex binds precursor proteins with reduced efficiency. We conclude that PE is required for the proper function of the TOM complex.
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, 10 mg/mL in chloroform, ≥97%, solution
Sigma-Aldrich
L-α-Phosphatidylethanolamine from Glycine max (soybean), Type IV, ≥98% (TLC), lyophilized powder
Sigma-Aldrich
L-α-Phosphatidylethanolamine from egg yolk, Type III, ≥97% (TLC), lyophilized powder