MilliporeSigma
  • Assessment of the genotoxic activity of alpha-asarone and its derivatives in the comet assay.
Assessment of the genotoxic activity of alpha-asarone and its derivatives in the comet assay.
Acta poloniae pharmaceutica (2013-04-26)
Jadwiga Marczewska, Ewa Drozd, Elzbieta Anuszewska, Zdzisław Chilmonczyk, Bozena Łozowicka
ABSTRACT
In our previous paper we examined mutagenic and genotoxic activity of 4 alpha-asarone isomers 2-5 exhibiting relatively high hypolipidemic activity. In the present paper, we examined genotoxic activity of alpha-asarone and its isomers as the ability to damage cellular DNA, evaluated in the comet assay. Additionally, mutagenic activity of alpha-asarone in Ames test has been examined. The Ames test for alpha-asarone was carried out in accordance with the guidelines of the PN-EN ISO 10993-3 standard. Compounds 4 and 5 were found to be devoid of any genotoxic activity while maintaining their hypolipemic potential. Because mutagenic activity of compound 4 was also minor it could be considered as a candidate for further pharmacological evaluation. Genotoxic but not mutagenic activity of alpha-asarone has been confirmed.