MilliporeSigma
  • Attenuation of portal hypertension by continuous portal infusion of PGE1 and immunologic impact in adult-to-adult living-donor liver transplantation.
Attenuation of portal hypertension by continuous portal infusion of PGE1 and immunologic impact in adult-to-adult living-donor liver transplantation.
Transplantation (2013-04-20)
Takashi Onoe, Yuka Tanaka, Kentaro Ide, Kouhei Ishiyama, Akihiko Oshita, Tsuyoshi Kobayashi, Hironobu Amano, Hirotaka Tashiro, Hideki Ohdan
ABSTRACT
Small-for-size syndrome remains the greatest limiting factor of expanding segmental liver transplantation from living donors. Portal hyperperfusion is considered to substantially contribute to small-for-size syndrome. We investigated the impact of continuous portal infusion of prostaglandin E1 (PGE1) on small-for-size grafts (SFSGs) in adult-to-adult living-donor liver transplantation (LDLT). From July 2003 to December 2009, LDLT was performed in 122 patients. We introduced continuous portal infusion of PGE1 to five SFSG patients (PG group) from November 2007 to December 2009 and retrospectively compared them with a historical control group of eight relevant SFSG patients without PGE1 infusion (non-PG group) from July 2003 to October 2007 to determine the safety and efficacy of continuous PGE1 portal infusion for SFSGs. Splenectomy cases were excluded from analysis. The PG group demonstrated significantly lower postoperative portal pressure than the non-PG group. Moreover, the PG group demonstrated significantly improved liver function in the early posttransplantation period and significantly better recovery from hyperammonemia at 1 week after transplantation and from hyperbilirubinemia in the late posttransplantation period. Overall survival was significantly better in the PG group than in the non-PG group. Three patients in the non-PG group died of rejection-related reasons. Interestingly, immunomonitoring assay revealed that antidonor immune responses were significantly accelerated in the non-PG group compared with the PG group after LDLT. In contrast, the PG group showed well-suppressed antidonor immune responses. Continuous portal infusion of PGE1 for SFSG attenuated portal hypertension, improved graft function, and suppressed antidonor immune responses, resulting in better survival.
Sigma-Aldrich
Prostaglandin E1, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, synthetic, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Prostaglandin E1, ≥98% (HPLC), synthetic
Alprostadil, European Pharmacopoeia (EP) Reference Standard