MilliporeSigma
  • Molecular view of an electron transfer process essential for iron-sulfur protein biogenesis.
Molecular view of an electron transfer process essential for iron-sulfur protein biogenesis.
Proceedings of the National Academy of Sciences of the United States of America (2013-04-19)
Lucia Banci, Ivano Bertini, Vito Calderone, Simone Ciofi-Baffoni, Andrea Giachetti, Deepa Jaiswal, Maciej Mikolajczyk, Mario Piccioli, Julia Winkelmann
ABSTRACT
Biogenesis of iron-sulfur cluster proteins is a highly regulated process that requires complex protein machineries. In the cytosolic iron-sulfur protein assembly machinery, two human key proteins--NADPH-dependent diflavin oxidoreductase 1 (Ndor1) and anamorsin--form a stable complex in vivo that was proposed to provide electrons for assembling cytosolic iron-sulfur cluster proteins. The Ndor1-anamorsin interaction was also suggested to be implicated in the regulation of cell survival/death mechanisms. In the present work we unravel the molecular basis of recognition between Ndor1 and anamorsin and of the electron transfer process. This is based on the structural characterization of the two partner proteins, the investigation of the electron transfer process, and the identification of those protein regions involved in complex formation and those involved in electron transfer. We found that an unstructured region of anamorsin is essential for the formation of a specific and stable protein complex with Ndor1, whereas the C-terminal region of anamorsin, containing the [2Fe-2S] redox center, transiently interacts through complementary charged residues with the FMN-binding site region of Ndor1 to perform electron transfer. Our results propose a molecular model of the electron transfer process that is crucial for understanding the functional role of this interaction in human cells.
Product Description
Riboflavin sodium phosphate, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, 73-79% (fluorimetric)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, synthetic, ≥70% (HPLC)
Sigma-Aldrich
Riboflavin 5′-monophosphate sodium salt hydrate, suitable for electrophoresis, suitable for acrylamide photopolymerization, ≥70%, powder
Sigma-Aldrich
Riboflavin Sodium Phosphate Hydrate, tested according to Ph. Eur.