MilliporeSigma
  • Effects of head down tilt on episcleral venous pressure in a rabbit model.
Effects of head down tilt on episcleral venous pressure in a rabbit model.
Experimental eye research (2013-04-10)
W J Lavery, J W Kiel
ABSTRACT
In humans, changing from upright to supine elicits an approximately 10 mmHg increase in cephalic venous pressure caused by the hydrostatic column effect, but episcleral venous pressure (EVP) and intraocular pressure (IOP) rise by only a few mmHg. The dissociation of the small increases in IOP and EVP compared to the larger increase in cephalic venous pressure suggests a regulatory mechanism controlling EVP. The aim of the present study was to determine if the rabbit model is suitable to study the effects of postural changes on EVP despite its short hydrostatic column. In anesthetized rabbits (n = 43), we measured arterial pressure (AP), IOP, and orbital venous pressure (OVP) by direct cannulation; carotid blood flow (BFcar) by transit time ultrasound, heart rate (HR) by digital cardiotachometer, and EVP with a servonull micropressure system. The goal of the protocol was to obtain measurement of supine EVP for ≈10 min, followed by ≈10 min of EVP measurement with the rabbit in a head down tilt. The data were analyzed by paired t-tests and the results reported as the mean ± standard error of the mean. In a separate group of animals (n = 35), aqueous flow was measured by fluorophotometry. This protocol entailed measurement of aqueous flow in the supine position for ≈60 min, followed by ≈60 min of aqueous flow measurement with the rabbit in a head down tilt. From supine to head down tilt, AP and BFcar were unchanged, IOP increased by 2.3 ± 0.4 mmHg (p < 0.001), EVP increased by 2.4 ± 0.4 mmHg (p < 0.001), OVP increased by 2.5 ± 0.2 mmHg (p < 0.001) and HR decreased by 9 ± 3 bpm (p = 0.002). Head down tilt caused no significant change in aqueous flow. Although the hydrostatic column in the rabbit is shorter than humans, the rabbit model permits sufficiently sensitive measurements of the pressures and systemic parameters likely involved in the EVP responses to posture change. The present results indicate directionally similar EVP and IOP responses to tilt as occur in humans and, as in humans, the responses are smaller than would be expected from the change in the hydrostatic column height. Also, as in humans, the model reveals no change in aqueous flow during head down tilt. We conclude the rabbit model is appropriate for studying the mechanisms responsible for the relative immunity of EVP and IOP to posture change.
Sigma-Aldrich
Hexamethonium hydroxide solution, ~0.1 M in H2O
Sigma-Aldrich
Hexamethonium bromide
Sigma-Aldrich
Hexamethonium chloride, solid (hygroscopic)