MilliporeSigma
  • Training health workers for magnesium sulfate use reduces case fatality from eclampsia: results from a multicenter trial.
Training health workers for magnesium sulfate use reduces case fatality from eclampsia: results from a multicenter trial.
Acta obstetricia et gynecologica Scandinavica (2013-04-05)
Friday E Okonofua, Rosemary N Ogu, Adetokunbo O Fabamwo, Innocent O Ujah, Calvin M Chama, Eric I Archibong, Hyacinth E Onah, Hadiza S Galadanci, James T Akuse
ABSTRACT
To investigate the effectiveness of an intervention aimed at improving the case management of eclampsia. A multi-center intervention study. Six teaching hospitals in Nigeria. Clinical records of cases of eclampsia treated before and 1 year after the intervention. Doctors and midwives in selected hospitals were re-trained to manage eclampsia using magnesium sulfate according to the Pritchard protocol. Eclampsia case fatality rates, maternal and perinatal mortality rates before and after the intervention. A total of 219 cases of eclampsia were managed over a 12-month period. There were seven maternal deaths. The post intervention case fatality rate of 3.2% was significantly less than the pre-intervention rate of 15.1% (p < 0.001). The overall maternal and perinatal mortality ratios and rates respectively in the hospitals declined from 1199.2 to 954 per 100,000 deliveries and 141.5 to 129.8 per 1000 births, respectively (p > 0.05). An intervention to build the capacity of care-providers to use an evidence-based protocol for the treatment of eclampsia in Nigeria was successful in reducing associated case fatality rate. The increased and widespread use of such an intervention in maternity units might contribute to the reduction of maternal mortality in low-income countries.
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, for molecular biology
Sigma-Aldrich
Magnesium sulfate solution, for molecular biology, 1.00 M±0.04 M
Sigma-Aldrich
Magnesium sulfate, anhydrous, reagent grade, ≥97%
Sigma-Aldrich
Magnesium sulfate, anhydrous, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate, puriss. p.a., drying agent, anhydrous, ≥98.0% (KT), powder (very fine)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioReagent, for molecular biology, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, ACS reagent, ≥98%
Sigma-Aldrich
Magnesium sulfate heptahydrate, puriss. p.a., ACS reagent, ≥99.0% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioXtra, ≥99.0%
Sigma-Aldrich
Magnesium sulfate heptahydrate, BioUltra, ≥99.5% (KT)
Sigma-Aldrich
Magnesium sulfate heptahydrate, 99.5-100.5% (calc. to the dried substance), meets analytical specification of Ph. Eur., BP,USP, FCC
Sigma-Aldrich
Magnesium sulfate heptahydrate, ReagentPlus®, ≥99.0%