MilliporeSigma
  • GLP-1 receptor activation and Epac2 link atrial natriuretic peptide secretion to control of blood pressure.
GLP-1 receptor activation and Epac2 link atrial natriuretic peptide secretion to control of blood pressure.
Nature medicine (2013-04-02)
Minsuk Kim, Mathew J Platt, Tadao Shibasaki, Susan E Quaggin, Peter H Backx, Susumu Seino, Jeremy A Simpson, Daniel J Drucker
ABSTRACT
Glucagon-like peptide-1 receptor (GLP-1R) agonists exert antihypertensive actions through incompletely understood mechanisms. Here we demonstrate that cardiac Glp1r expression is localized to cardiac atria and that GLP-1R activation promotes the secretion of atrial natriuretic peptide (ANP) and a reduction of blood pressure. Consistent with an indirect ANP-dependent mechanism for the antihypertensive effects of GLP-1R activation, the GLP-1R agonist liraglutide did not directly increase the amount of cyclic GMP (cGMP) or relax preconstricted aortic rings; however, conditioned medium from liraglutide-treated hearts relaxed aortic rings in an endothelium-independent, GLP-1R-dependent manner. Liraglutide did not induce ANP secretion, vasorelaxation or lower blood pressure in Glp1r(-/-) or Nppa(-/-) mice. Cardiomyocyte GLP-1R activation promoted the translocation of the Rap guanine nucleotide exchange factor Epac2 (also known as Rapgef4) to the membrane, whereas Epac2 deficiency eliminated GLP-1R-dependent stimulation of ANP secretion. Plasma ANP concentrations were increased after refeeding in wild-type but not Glp1r(-/-) mice, and liraglutide increased urine sodium excretion in wild-type but not Nppa(-/-) mice. These findings define a gut-heart GLP-1R-dependent and ANP-dependent axis that regulates blood pressure.
Sigma-Aldrich
Natriuretic Peptide, C-Type, ≥95% (HPLC)
Sigma-Aldrich
Guanosine 3′,5′-cyclic monophosphate, ≥98% (HPLC), powder