MilliporeSigma
  • Effectiveness of lurasidone for patients with schizophrenia following 6 weeks of acute treatment with lurasidone, olanzapine, or placebo: a 6-month, open-label, extension study.
Effectiveness of lurasidone for patients with schizophrenia following 6 weeks of acute treatment with lurasidone, olanzapine, or placebo: a 6-month, open-label, extension study.
The Journal of clinical psychiatry (2013-04-02)
Stephen M Stahl, Josephine Cucchiaro, Doreen Simonelli, Jay Hsu, Andrei Pikalov, Antony Loebel
ABSTRACT
The primary objective was to evaluate the safety and tolerability of lurasidone, a new atypical antipsychotic agent, in the longer-term treatment of schizophrenia (DSM-IV). Persistence of symptom improvement was assessed as a secondary outcome. Patients who completed a 6-week, double-blind, placebo-controlled study evaluating the efficacy of fixed doses of once-daily lurasidone (40 or 120 mg) or olanzapine 15 mg (to confirm assay sensitivity) were eligible to receive flexibly dosed lurasidone 40 to 120 mg/d in this 6-month, open-label extension study (conducted from March 2008 to December 2009). Assessments of safety and tolerability were conducted at open-label baseline, at day 10, and monthly thereafter. Of 254 enrolled patients, 113 (44.5%) completed 6 months of open-label treatment. During the open-label study (month 6 observed cases), small decreases were observed in mean weight (-0.1 kg) and median lipid levels (total cholesterol, -6.5 mg/dL; low-density lipoprotein, 0.0 mg/dL; high-density lipoprotein, 0.0 mg/dL; triglycerides, -8.5 mg/dL). Patients previously treated with olanzapine (n = 69) experienced decrease in weight and improvement in lipid levels, whereas patients previously treated with lurasidone (n = 115) or placebo (n = 62) experienced minimal changes. No clinically meaningful changes were observed in median prolactin levels. The 2 most commonly reported adverse events were akathisia (13.0%) and insomnia (11.0%). Persistent antipsychotic efficacy of lurasidone was shown for patients who had previously received lurasidone, olanzapine, or placebo; further reductions from open-label baseline to final visit were observed in mean PANSS total score (-8.7) for all patients. Open-label treatment with flexibly dosed lurasidone (40-120 mg/d) was generally safe, well tolerated, and effective over a 6-month period in patients who had completed a preceding 6-week, double-blind study.
Product Description
Supelco
Olanzapine solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Olanzapine, ≥98% (HPLC)