MilliporeSigma
α-Cyclodextrin decreases cholera toxin binding to GM1-gangliosides.
Journal of medical microbiology (2013-03-30)
Boris Ermolinsky, Michael Peredelchuk, Daniele Provenzano
ABSTRACT
Cholera toxin (CT), the principal virulence factor secreted by Vibrio cholerae, is an A-B5 type exotoxin that binds to host cell GM1-gangliosides and is responsible for cholera diarrhoea. We tested the hypothesis that the cyclic hexasaccharide α-cyclodextrin (α-CD), but not the cyclic heptasaccharides methyl-β-cyclodextrin (MD-β-CD) and hydroxypropyl-β-cyclodextrin (HP-β-CD) inhibit binding of CT to GM1-gangliosides. We report that α-CD decreases CT binding to GM1-ganglioside-coated microtitre plate wells and on the surface of fixed HeLa cells in a concentration-dependent manner, suggesting that this may be a promising lead for the development of compounds with therapeutic properties.
Sigma-Aldrich
α-Cyclodextrin, ≥98%
Sigma-Aldrich
α-Cyclodextrin, powder, BioReagent, suitable for cell culture, ≥98%
Sigma-Aldrich
α-Cyclodextrin, purum, ≥98.0% (HPLC)
Sigma-Aldrich
α-Cyclodextrin, Produced by Wacker Chemie AG, Burghausen, Germany, Life Science, 98.0-101.0% cyclodextrin basis (HPLC)
Sigma-Aldrich
α-Cyclodextrin, produced by Wacker Chemie AG, Burghausen, Germany, ≥98.0% (HPLC)