MilliporeSigma
  • Loss of sorting nexin 27 contributes to excitatory synaptic dysfunction by modulating glutamate receptor recycling in Down's syndrome.
Loss of sorting nexin 27 contributes to excitatory synaptic dysfunction by modulating glutamate receptor recycling in Down's syndrome.
Nature medicine (2013-03-26)
Xin Wang, Yingjun Zhao, Xiaofei Zhang, Hedieh Badie, Ying Zhou, Yangling Mu, Li Shen Loo, Lei Cai, Robert C Thompson, Bo Yang, Yaomin Chen, Peter F Johnson, Chengbiao Wu, Guojun Bu, William C Mobley, Dongxian Zhang, Fred H Gage, Barbara Ranscht, Yun-wu Zhang, Stuart A Lipton, Wanjin Hong, Huaxi Xu
ABSTRACT
Sorting nexin 27 (SNX27), a brain-enriched PDZ domain protein, regulates endocytic sorting and trafficking. Here we show that Snx27(-/-) mice have severe neuronal deficits in the hippocampus and cortex. Although Snx27(+/-) mice have grossly normal neuroanatomy, we found defects in synaptic function, learning and memory and a reduction in the amounts of ionotropic glutamate receptors (NMDA and AMPA receptors) in these mice. SNX27 interacts with these receptors through its PDZ domain, regulating their recycling to the plasma membrane. We demonstrate a concomitant reduced expression of SNX27 and CCAAT/enhancer binding protein β (C/EBPβ) in Down's syndrome brains and identify C/EBPβ as a transcription factor for SNX27. Down's syndrome causes overexpression of miR-155, a chromosome 21-encoded microRNA that negatively regulates C/EBPβ, thereby reducing SNX27 expression and resulting in synaptic dysfunction. Upregulating SNX27 in the hippocampus of Down's syndrome mice rescues synaptic and cognitive deficits. Our identification of the role of SNX27 in synaptic function establishes a new molecular mechanism of Down's syndrome pathogenesis.
Sigma-Aldrich
Ifosfamide, ≥98%
Pricing and availability is not currently available.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, meets USP testing specifications
Pricing and availability is not currently available.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Mitomycin C from Streptomyces caespitosus, powder, contains NaCl as solubilizer
Pricing and availability is not currently available.