MilliporeSigma
  • Pseudo-first-order alkaline hydrolysis of diethyl tartrate: a baseline study for a polymer matrix used in controlled-release delivery systems.
Pseudo-first-order alkaline hydrolysis of diethyl tartrate: a baseline study for a polymer matrix used in controlled-release delivery systems.
Journal of pharmaceutical sciences (1990-04-01)
D S Kalonia, A P Simonelli
ABSTRACT
The hydrolysis kinetics of a bifunctional group compound, diethyl tartrate, was studied as a function of temperature and pH in the alkaline region. A pH-stat was used to maintain constant pH conditions in the alkaline region. This allowed the studies to be carried out at low ionic strengths and without the use of buffers. The results indicate that the hydrolysis for both steps followed specific base catalysis. The ratio of the two rate constants was 13.31, which was attributed to a strong charge effect in the second step. The results also show that the use of an overall average rate constant may not be acceptable for multifunctional group compounds.
Sigma-Aldrich
(+)-Diethyl L-tartrate, ≥99%
Sigma-Aldrich
Diethyl L-tartrate, ≥99%, FG
Sigma-Aldrich
(−)-Diethyl D-tartrate, ≥99%