MilliporeSigma
  • Comprehensive analysis of the transcription of starch synthesis genes and the transcription factor RSR1 in wheat (Triticum aestivum) endosperm.
Comprehensive analysis of the transcription of starch synthesis genes and the transcription factor RSR1 in wheat (Triticum aestivum) endosperm.
Genome (2013-03-23)
Guo-Zhang Kang, Wei Xu, Guo-Qin Liu, Xiao-Qi Peng, Tian-Cai Guo
ABSTRACT
The cDNA sequences of 26 starch synthesis genes were identified in common wheat (Triticum aestivum L.), and their transcript levels were measured using quantitative real-time RT-PCR to assess the function of individual genes and the regulatory mechanism in wheat endosperm. The expression patterns of 26 genes in wheat endosperm were classified into three groups. The genes in group 1 were richly expressed in the early stage of grain development and may be involved in the construction of fundamental cell machinery, synthesis of glucan primers, and initiation of starch granules. The genes in group 2 were highly expressed during the middle and late stages of grain development, and their expression profiles were similar to the accumulation rate of endosperm starch; these genes are presumed to play a crucial role in starch production. The genes in group 3 were scantily expressed throughout the grain development period and might be associated with transitory starch synthesis. Transcripts of the negative transcription factor TaRSR1 were high at the early and late stages of grain development but low during the middle stage. The expression pattern of TaRSR1 was almost opposite to those of the group 2 starch synthesis genes, indicating that TaRSR1 might negatively regulate the expression of many endosperm starch synthesis genes during grain development.
Sigma-Aldrich
Starch from wheat, Unmodified
Sigma-Aldrich
Starch from potato, suitable for electrophoresis
Sigma-Aldrich
Starch from potato, Powder
Sigma-Aldrich
Starch from potato, Soluble
Sigma-Aldrich
Starch, from potato, tested according to Ph. Eur.
Sigma-Aldrich
Starch, puriss. p.a., from potato, reag. ISO, reag. Ph. Eur., soluble
Sigma-Aldrich
Starch from corn
Supelco
Starch from corn, analytical standard, analytical standard for Starch Assay Kits SA-20 and STA-20
Sigma-Aldrich
Starch from corn
Sigma-Aldrich
Starch from corn, practical grade
Sigma-Aldrich
Starch from rice
Supelco
Starch from corn, for use with Total Dietary Fiber Control Kit, TDF-C10