MilliporeSigma
  • Essential oil of flowers of Anaphalis contorta, an aromatic and medicinal plant from India.
Essential oil of flowers of Anaphalis contorta, an aromatic and medicinal plant from India.
Natural product communications (2013-03-22)
Rajesh K Joshi
ABSTRACT
The hydro-distilled essential oil of dried flowers of Anaphalis contorta Hook f. has been examined by means of gas chromatography-mass spectrometry (GC-MS). The oil constituents were identified according to their mass spectra and their relative retention indices determined on a non-polar stationary phase capillary column. Fifty-one constituents have been identified representing 97.0% of the total oil. The major constituents were beta-caryophyllene (19.2%), gamma-curcumene (17.5%), delta-cadinene (10.2%), labda-7,14-dien-13-ol (4.8%), epi-alpha-cadinol (4.3%), bulnesol (4.3%), alpha-cadinol (3.8%), beta-bisabolol (3.7%) and labda-8,14-dien-13-ol (3.3%). The oil was rich in sesquiterpene hydrocarbons (60.5%), followed by oxygenated sesquiterpenes (26.6%), oxygenated diterpenes (8.1%) and diterpene hydrocarbons (1.8%).
Sigma-Aldrich
(−)-trans-Caryophyllene, ≥98.0% (sum of enantiomers, GC)
Supelco
(−)-trans-Caryophyllene, analytical standard
Sigma-Aldrich
β-Caryophyllene, ≥80%, FCC, FG