MilliporeSigma
  • Case report: Crohn's-like mycophenolate-induced colitis, a fallout in steroid-free regimens.
Case report: Crohn's-like mycophenolate-induced colitis, a fallout in steroid-free regimens.
Transplantation proceedings (2013-03-19)
A D Jakes, A Roy, M Veerasamy, S Bhandari
ABSTRACT
Diarrhea is considered to be a common side effect after renal transplantation, due to a number of infective or drug-related causes. Over the past decade the etiology has perhaps changed with the evolution of immunosuppression. In an attempt to minimize early acute rejection and potential steroid use, the recent introduction of mycophenolic acid-based therapies has increased the incidence of diarrheal symptoms. Histologic and macroscopic appearance of mycophenolate-induced colitis is not well documented. Three patients with immunosuppression-induced colitis had received deceased-donor renal transplantations and presented with diarrhea and/or abdominal pain. All patients made a full recovery after decreasing the dose or withdrawing mycophenolate mofetil or myfortic with corticosteroid-free regimens. Patients with immunosuppression-induced colitis require prompt intervention by dose reduction or withdrawal. Both myocophenolate mofetil and myfortic can induce a Crohn's-like colitis even after a long period of exposure. The symptoms may require 6 months to resolve, suggesting that surgery should be considered only as a last resort after a significant period off therapy.
Product Description
Supelco
Mycophenolic acid solution, 1.0 mg/mL in acetonitrile, ampule of 1 mL, certified reference material, Cerilliant®
Sigma-Aldrich
Mycophenolic acid, ≥98%
Sigma-Aldrich
Mycophenolic acid, powder, BioReagent, suitable for cell culture