MilliporeSigma
  • Fabrication and characterization of bioactive β-Ca2SiO4/PHBV composite scaffolds.
Fabrication and characterization of bioactive β-Ca2SiO4/PHBV composite scaffolds.
Materials science & engineering. C, Materials for biological applications (2013-03-19)
Nana Wang, Zheng Zhou, Leilei Xia, Yao Dai, Hairong Liu
ABSTRACT
A key challenge in tissue engineering is the construction of a scaffold with adequate properties which would mimic extracellular matrix (ECM) to induce the cells' efficient adhesion, proliferation and proper differentiation. Novel β-Ca2SiO4/PHBV composite scaffolds were fabricated by integrating β-Ca2SiO4 nanoparticles with PHBV backbone via a modified solvent casting-particulates leaching method, which generates interconnected porous structure and the high porosity, about 87%, of these scaffolds. Compared with PHBV scaffolds, β-Ca2SiO4/PHBV composite scaffolds facilitate the adhesion of human osteoblast-like MG-63 cells due to their increased hydrophilicity. The β-Ca2SiO4/PHBV composite scaffolds containing 2.5 or 5% β-Ca2SiO4 nanoparticles significantly enhance the proliferation of MG-63 cells by stimulating the transcription of the transforming growth factor-β1 (TGF-β1) and bone morphogenetic protein-7 (BMP-7) genes. These scaffolds also induce early differentiation via promoting the transcription of alkaline phosphatase (ALP). The results suggest the potential application of β-Ca2SiO4/PHBV composites in bone tissue engineering.
Sigma-Aldrich
Calcium silicate, purum, 12-22% Ca (as CaO) basis, ≥87% SiO2 basis
Sigma-Aldrich
Calcium silicate, −200 mesh, 99%