MilliporeSigma
  • Isolation of Shigellae. 8. Comparison of xylose lysine deoxycholate agar, hektoen enteric agar, Salmonella-Shigella agar, and eosin methylene blue agar with stool specimens.
Isolation of Shigellae. 8. Comparison of xylose lysine deoxycholate agar, hektoen enteric agar, Salmonella-Shigella agar, and eosin methylene blue agar with stool specimens.
Applied microbiology (1971-01-01)
W I Taylor, D Schelhart
ABSTRACT
Two enrichment broths and four plating media were compared for efficiency of detection of enteric pathogens from 1,597 stool specimens. Of 170 salmonellae isolated from the composite of all methods, direct streaking yielded but 54%, whereas enrichment in gram-negative broth found 87% and Selenite-F broth 97%. By contrast, gram-negative broth produced 100% of the 17 shigellae, Selenite-F broth but 77%, and direct streaking only 59%. Thus, enrichment methods produced almost twice the number of both pathogens as direct streaking. Comparison of the plating media revealed xylose lysine deoxycholate agar (XLD) and Hektoen enteric agar to be equal in their abilities to find both pathogens. Both were moderately better than Salmonella-Shigella agar and markedly superior to eosin methylene blue agar. XLD fround 83% of salmonellae produced by the composite of four media and 90% of the shigellae. Hektoen enteric agar found 80% of both. Salmonella-Shigella agar detected 74 and 68%, respectively, and eosin methylene blue agar only 42 and 63%. The numbers of false positives accruing to each medium, however, showed Hektoen enteric and Salmonella-Shigella agars to produce more than twice as many false-positive plates as XLD. Similarly, Selenite-F broth resulted in many more false-positives for all plating media than did gram-negative broth. Consequently, the index of validity, which equates successful isolation of pathogens with total pickings, favored XLD and gram-negative broth as the media of choice, with direct streaking the poorest method by all counts.
Product Description
Millipore
Hektoen Enteric Agar, NutriSelect® Plus, powder, selective and differential for Salmonella spp., for Shigella spp., pack of 500 g, suitable for microbiology