MilliporeSigma
  • Permeation of insulin, calcitonin and exenatide across Caco-2 monolayers: measurement using a rapid, 3-day system.
Permeation of insulin, calcitonin and exenatide across Caco-2 monolayers: measurement using a rapid, 3-day system.
PloS one (2013-03-14)
Vivek Gupta, Nishit Doshi, Samir Mitragotri
ABSTRACT
Caco-2 monolayers are one of the most widely used in vitro models for prediction of intestinal permeability of therapeutic molecules. However, the conventional Caco-2 monolayer model has several drawbacks including labor-intensive culture process, unphysiological growth conditions, lack of reproducibility and limited throughput. Here, we report on the use of 3-day Caco-2 monolayers for assessing permeability of polypeptide drugs. The 3-day monolayers were grown in a commercially available transwell set-up, which facilitates rapid development of the Caco-2 monolayers in an intestinal epithelial differentiation mimicking environment. This set-up included use of serum-free medium of defined composition with supplements such as butyric acid, hormones, growth factors, and other metabolites, reported to regulate the differentiation of intestinal epithelial cells in vivo. We measured permeability of 3 different therapeutic polypeptides; insulin, calcitonin, and exenatide across the monolayer. Preliminary validation of the monolayer was carried out by confirming dose-dependent permeation of FITC-insulin and sulforhodamine-B. Transport of insulin, calcitonin, and exenatide measured at different loading concentrations suggests that the permeability values obtained with 3-day cultures resemble more closely the values obtained with ex vivo models compared to permeability values obtained with conventional 21-day cultures. Short-term 3-day Caco-2 monolayers provide new opportunities for developing reproducible and high-throughput models for screening of therapeutic macromolecules for oral absorption.
Sigma-Aldrich
Sulforhodamine B, Dye content 75 %
Sigma-Aldrich
Sulforhodamine B, acid form, Dye content 95 %
Sigma-Aldrich
Sulforhodamine B sodium salt, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
Sulforhodamine B sodium salt, Technical grade