MilliporeSigma
  • Antagonism of a zinc metalloprotease using a unique metal-chelating scaffold: tropolones as inhibitors of P. aeruginosa elastase.
Antagonism of a zinc metalloprotease using a unique metal-chelating scaffold: tropolones as inhibitors of P. aeruginosa elastase.
Chemical communications (Cambridge, England) (2013-03-14)
Jessica L Fullagar, Amanda L Garner, Anjali K Struss, Joshua A Day, David P Martin, Jing Yu, Xiaoqing Cai, Kim D Janda, Seth M Cohen
ABSTRACT
Tropolone emerged from the screening of a chelator fragment library (CFL) as an inhibitor of the Zn(2+)-dependent virulence factor, Pseudomonas aeruginosa elastase (LasB). Based on this initial hit, a series of substituted tropolone-based LasB inhibitors was prepared, and a compound displaying potent activity in vitro and in a bacterial swarming assay was identified. Importantly, this inhibitor was found to be specific for LasB over other metalloenzymes, validating the usage of tropolone as a viable scaffold for identifying first-in-class LasB inhibitors.
Sigma-Aldrich
Tropolone, 98%