MilliporeSigma
  • Synthesis and characterization of the in situ bulk polymerization of PMMA containing graphene sheets using microwave irradiation.
Synthesis and characterization of the in situ bulk polymerization of PMMA containing graphene sheets using microwave irradiation.
Molecules (Basel, Switzerland) (2013-03-13)
Mohammad A Aldosari, Ali A Othman, Edreese H Alsharaeh
ABSTRACT
Polymethylmethacrylate-graphene (PMMA/RGO) nanocomposites were prepared via in situ bulk polymerization using two different preparation techniques. In the first approach, a mixture of graphite oxide (GO) and methylmethacrylate monomers (MMA) were polymerized using a bulk polymerization method with a free radical initiator. After the addition of the reducing agent hydrazine hydrate (HH), the product was reduced via microwave irradiation (MWI) to obtain R-(GO-PMMA) composites. In the second approach, a mixture of graphite sheets (RGO) and MMA monomers were polymerized using a bulk polymerization method with a free radical initiator to obtain RGO-(PMMA) composites. The composites were characterized by FTIR, (1)H-NMR and Raman spectroscopy and XRD, SEM, TEM, TGA and DSC. The results indicate that the composite obtained using the first approach, which involved MWI, had a better morphology and dispersion with enhanced thermal stability compared with the composites prepared without MWI.
Sigma-Aldrich
Poly(methyl methacrylate)
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~350,000 by GPC
Sigma-Aldrich
Poly(methyl methacrylate)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,480,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 8,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 50,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 20,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 100,000
Sigma-Aldrich
Poly(methyl methacrylate), average Mw ~15,000 by GPC, powder
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 2,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 10,000
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, average Mw 97,000 (Typical), average Mn 46,000 (Typical)
Supelco
Poly(methyl methacrylate), analytical standard, for GPC, 4,000
Sigma-Aldrich
Poly(methyl methacrylate), isotactic, >80% isotactic