MilliporeSigma
  • Biochemical correlates of neuropsychiatric illness in maple syrup urine disease.
Biochemical correlates of neuropsychiatric illness in maple syrup urine disease.
The Journal of clinical investigation (2013-03-13)
Emilie R Muelly, Gregory J Moore, Scott C Bunce, Julie Mack, Don C Bigler, D Holmes Morton, Kevin A Strauss
ABSTRACT
Maple syrup urine disease (MSUD) is an inherited disorder of branched chain amino acid metabolism presenting with neonatal encephalopathy, episodic metabolic decompensation, and chronic amino acid imbalances. Dietary management enables survival and reduces risk of acute crises. Liver transplantation has emerged as an effective way to eliminate acute decompensation risk. Psychiatric illness is a reported MSUD complication, but has not been well characterized and remains poorly understood. We report the prevalence and characteristics of neuropsychiatric problems among 37 classical MSUD patients (ages 5-35 years, 26 on dietary therapy, 11 after liver transplantation) and explore their underlying mechanisms. Compared with 26 age-matched controls, MSUD patients were at higher risk for disorders of cognition, attention, and mood. Using quantitative proton magnetic resonance spectroscopy, we found lower brain glutamate, N-acetylaspartate (NAA), and creatine concentrations in MSUD patients, which correlated with specific neuropsychiatric outcomes. Asymptomatic neonatal course and stringent longitudinal biochemical control proved fundamental to optimizing long-term mental health. Neuropsychiatric morbidity and neurochemistry were similar among transplanted and nontransplanted MSUD patients. In conclusion, amino acid dysregulation results in aberrant neural networks with neurochemical deficiencies that persist after transplant and correlate with neuropsychiatric morbidities. These findings may provide insight into general mechanisms of psychiatric illness.
Product Description
Supelco
L-Aspartic acid, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
L-Aspartic acid, BioXtra, ≥99% (HPLC)
Sigma-Aldrich
L-Aspartic acid hemimagnesium salt dihydrate, ≥97.0% (KT)
Sigma-Aldrich
L-Aspartic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
N-Acetyl-L-aspartic acid, ≥99.0% (T)
SAFC
L-Aspartic acid
Supelco
L-Aspartic acid, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Aspartic acid, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Aspartic acid, from non-animal source, meets EP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
DL-Aspartic acid, ≥99% (TLC)
Sigma-Aldrich
Creatine, anhydrous
Sigma-Aldrich
L-Aspartic acid potassium salt, ≥98% (HPLC)