MilliporeSigma
  • Steroids glycosylated with both D- and L-arabinoses from the South China Sea gorgonian Dichotella gemmacea.
Steroids glycosylated with both D- and L-arabinoses from the South China Sea gorgonian Dichotella gemmacea.
Journal of natural products (2013-03-13)
Mei Jiang, Peng Sun, Hua Tang, Bao-Shu Liu, Tie-Jun Li, Cui Li, Wen Zhang
ABSTRACT
Three new 19-hydroxy steroidal glycosides, namely, junceellosides E-G (2-4), were isolated together with the known analogue junceelloside C (1) from the South China Sea gorgonian Dichotella gemmacea. The structures of these compounds were elucidated by a combination of detailed spectroscopic analyses, chemical methods, and comparison with reported data. These glycosides are found to have sugar moieties of both β-l- and β-d-arabinopyranoses by HPLC analysis of their thiocarbamoyl-thiazolidine derivatives and those of authentic d- and l-arabinoses, leading to the structure revision of junceelloside C (1). This is the first report of steroidal glycosides from the gorgonian D. gemmacea and the first report of glycosides with β-l-arabinopyranose from marine sources.
Sigma-Aldrich
DL-Arabinose, ≥98% (GC)