MilliporeSigma
  • Chronic pulmonary dysfunction following acute inhalation of butyl acrylate.
Chronic pulmonary dysfunction following acute inhalation of butyl acrylate.
The West Virginia medical journal (2013-03-12)
Ravindra Bhardwaj, Alan Ducatman, Mitchell S Finkel, Edward Petsonk, Janet Hunt, Robert J Beto
ABSTRACT
Butyl Acrylate (BA) (2-propionic acid; CH2 = CHCOOC4H9) is a colorless liquid commonly used in impregnation agents and adhesives. Dermal contact with BA has previously been reported to cause moderate skin irritation with skin sensitizing potential in humans. Health effects of inhalation of BA have not been previously reported. Accordingly, we document the health conditions of a bystander, first responder and landfill worker exposed to butyl acrylate (BA) released to the atmosphere following a collision and roadside spill in October 1998. Retrospective data were collected via chart review and analyzed for exposure, symptoms, physical findings and radiological, laboratory and spirometry results over a ten-year period. All three patients had similar respiratory symptoms including a dramatic hacking cough and dyspnea. Findings included abnormal pulmonary function tests and breath sounds. These data underscore the potential hazards of BA inhalational exposure and the need to wear additional protective equipment.
Product Description
Supelco
Butyl acrylate, analytical standard
Sigma-Aldrich
Butyl acrylate, ≥99%, contains 10-60 ppm monomethyl ether hydroquinone as inhibitor