MilliporeSigma
[Tissue engineering using porous polyethylene implants].
HNO (2013-03-08)
S Strieth
ABSTRACT
Although porous polyethylene (PPE) implants are already used in reconstructive craniofacial surgery, they are still only applied in rhinoplasty when critically indicated. Biocompatibility of PPE implants (pore size ~100-200 µm) can be modified by tissue engineering: coatings with extracellular matrix components, growth factors, anti-inflammatory substances and living cells are all possible. At present, transparent dorsal skinfold chamber models in animals appear to be particularly appropriate for pre-clinical evaluation. Using in vivo fluorescence microscopy, angiogenic microvascular ingrowth and inflammatory leukocyte-endothelial cell interactions can be analysed in detail with regard to functionally relevant mechanical integration. Preclinical data indicate that tissue engineering could improve the biocompatibility of these novel PPE implants and thus extend their range of clinical applications.
Sigma-Aldrich
Polyethylene, High density, melt index 2.2 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, High density, melt index 12 g/10 min (190 °C/2.16kg)
Sigma-Aldrich
Polyethylene, low density, melt index 25 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Linear low density, melt index 1.0 g/10 min (190°C/2.16kg)
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 34-50 μm particle size
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, average Mw 3,000,000-6,000,000
Sigma-Aldrich
Polyethylene, average Mw ~4,000 by GPC, average Mn ~1,700 by GPC
Sigma-Aldrich
Polyethylene, Ultra-high molecular weight, surface-modified, powder, 125 μm avg. part. size
Sigma-Aldrich
Polyethylene, Medium density
Supelco
Polyethylene, analytical standard, 2,000
Polyethylene (LDPE), ERM®, certified reference material