MilliporeSigma
  • Removal of airborne toxic chemicals by porous organic polymers containing metal-catecholates.
Removal of airborne toxic chemicals by porous organic polymers containing metal-catecholates.
Chemical communications (Cambridge, England) (2013-03-07)
Mitchell H Weston, Gregory W Peterson, Matthew A Browe, Paulette Jones, Omar K Farha, Joseph T Hupp, Sonbinh T Nguyen
ABSTRACT
Porous organic polymers bearing metal-catecholate groups were evaluated for their ability to remove airborne ammonia, cyanogen chloride, sulphur dioxide, and octane by micro-breakthrough analysis. For ammonia, the metal-catecholate materials showed remarkable uptake under humid conditions.
Sigma-Aldrich
Octane, electronic grade, ≥99.999% metals basis, ≥99% (CP)
Supelco
Octane, analytical standard
Sigma-Aldrich
Sulfur dioxide, ≥99.9%
Sigma-Aldrich
Octane, anhydrous, ≥99%
Sigma-Aldrich
Octane, reagent grade, 98%