MilliporeSigma
  • Gene cloning and characterization of MdeA, a novel multidrug efflux pump in Streptococcus mutans.
Gene cloning and characterization of MdeA, a novel multidrug efflux pump in Streptococcus mutans.
Journal of microbiology and biotechnology (2013-03-07)
Do Kyun Kim, Kyoung Hoon Kim, Eun Ji Cho, Seoung-Je Joo, Jung-Min Chung, Byoung Yil Son, Jong Hwa Yum, Young-Man Kim, Hyun-Ju Kwon, Byung-Woo Kim, Tae Hoon Kim, Eun-Woo Lee
ABSTRACT
Multidrug resistance, especially multidrug efflux mechanisms that extrude structurally unrelated cytotoxic compounds from the cell by multidrug transporters, is a serious problem and one of the main reasons for the failure of therapeutic treatment of infections by pathogenic microorganisms as well as of cancer cells. Streptococcus mutans is considered one of the primary causative agents of dental caries and periodontal disease, which comprise the most common oral diseases. A fragment of chromosomal DNA from S. mutans KCTC3065 was cloned using Escherichia coli KAM32 as host cells lacking major multidrug efflux pumps. Although E. coli KAM32 cells were very sensitive to many antimicrobial agents, the transformed cells harboring a recombinant plasmid became resistant to several structurally unrelated antimicrobial agents such as tetracycline, kanamycin, rhodamin 6G, ampicillin, acriflavine, ethidium bromide, and tetraphenylphosphonium chloride. This suggested that the cloned DNA fragment carries a gene encoding a multidrug efflux pump. Among 49 of the multidrug-resistant transformants, we report the functional gene cloning and characterization of the function of one multidrug efflux pump, namely MdeA from S. mutans, which was expressed in E. coli KAM32. Judging from the structural and biochemical properties, we concluded that MdeA is the first cloned and characterized multidrug efflux pump using the proton motive force as the energy for efflux drugs.
Sigma-Aldrich
Tetraphenylphosphonium bromide, 97%
Sigma-Aldrich
Tetraphenylphosphonium chloride, 98%