MilliporeSigma
  • Unprecedented one-pot, domino tertiary alcohol protection-Michael type addition of halides to Morita-Baylis-Hillman adduct of isatin with RCOX/K2CO3: diastereoselective synthesis of oxindole appended β-halo esters.
Unprecedented one-pot, domino tertiary alcohol protection-Michael type addition of halides to Morita-Baylis-Hillman adduct of isatin with RCOX/K2CO3: diastereoselective synthesis of oxindole appended β-halo esters.
Organic letters (2013-03-07)
Rajarethinam Solaiselvi, Ponnusamy Shanmugam, Asit Baran Mandal
ABSTRACT
A facile method utilizing RCOX/K2CO3 as a novel reagent for conjugate addition of hydrogen halide, in addition to tertiary (3°)-hydroxyl protection that leads to the synthesis of functionalized β-halo Morita-Baylis-Hillman ester appended oxindoles, has been developed. The diastereoselective one-pot O-acylation-hydrohalogenation observed cannot otherwise be performed by treatment with hydrohalide. Deprotection of a 3°-hydroxyl protecting group has also been demonstrated by treatment with hydrochloric acid.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, for molecular biology
Sigma-Aldrich
Isatin, 97%
Supelco
Hydrogen chloride – ethanol solution, ~1.25 M HCl, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid, puriss., 24.5-26.0%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, puriss. p.a., ACS reagent, reag. ISO, reag. Ph. Eur., fuming, ≥37%, APHA: ≤10
Supelco
Hydrogen chloride - 1-butanol solution, ~3 M in 1-butanol, for GC derivatization, LiChropur
Sigma-Aldrich
Hydrogen chloride, ReagentPlus®, ≥99%