MilliporeSigma
  • Crotonaldehyde-exposed macrophages induce IL-8 release from airway epithelial cells through NF-κB and AP-1 pathways.
Crotonaldehyde-exposed macrophages induce IL-8 release from airway epithelial cells through NF-κB and AP-1 pathways.
Toxicology letters (2013-03-06)
Bi-Cheng Yang, Zhi-Hua Yang, Xiu-Jie Pan, Feng-Jun Xiao, Xing-Yu Liu, Mao-Xiang Zhu, Jian-Ping Xie
ABSTRACT
Crotonaldehyde, a highly toxic α, β-unsaturated aldehyde, is a major component of cigarette smoke and a ubiquitous environmental pollutant. Crotonaldehyde exposure is known to have adverse effects on respiratory health, but the underlying mechanisms remain obscure. To examine the interaction between macrophages and airway epithelial cells after exposure to crotonaldehyde, BEAS-2B and A549 cells were treated with conditioned media from a human monocytic leukemia cell line (THP-1) cells stimulated with crotonaldehyde. We demonstrate that conditioned media from THP-1 cells stimulated with crotonaldehyde increased interleukin (IL)-8 production, enhanced nuclear factor (NF)-κB and AP-1 DNA-binding activity in BEAS-2B and A549 cells. Analysis of these conditioned media revealed marked increases in tumor necrosis factor (TNF)-α, IL-1β and IL-8 levels. Preincubation of conditioned media with either TNF-α- or IL-1β-neutralizing antibodies reduced IL-8 production. Furthermore, BEAS-2B and A549 cells directly treated with crotonaldehyde induced increase in IL-8 production. These data suggest that crotonaldehyde is capable of directly stimulating the production of IL-8 in both macrophages and airway epithelial cells. Crotonaldehyde-stimulated macrophages also amplify the inflammatory response by enhancing IL-8 release from airway epithelial cells mediated by NF-κB and AP-1 pathways through a mechanism involving TNF-α and IL-1β. These findings indicate that crotonaldehyde can cause lung inflammatory response via multiple mechanisms, and may contribute to chronic airway inflammation in smokers.
Sigma-Aldrich
Crotonaldehyde, predominantly trans, ≥99%, contains 0.1-0.2% BHT as stabilizer, 1% H2O as stabilizer
Sigma-Aldrich
Crotonaldehyde, mixture of cis and trans, ratio of cis- and trans-isomers (~1:20), ≥99.5% (GC)