MilliporeSigma
  • Design and development of insulin emulgel formulation for transdermal drug delivery and its evaluation.
Design and development of insulin emulgel formulation for transdermal drug delivery and its evaluation.
Pakistan journal of pharmaceutical sciences (2013-03-05)
Muhammad Akram, Syed Baqirshyum Naqvi, Ahmad Khan
ABSTRACT
The objective of the present study was to formulate an insulin emulgel, selection of an optimize formulation through in vitro drug release kinetics and finally evaluate its hypoglycemic activity in animal model. Insulin emulgel was prepared using emu oil as penetration enhancer with the combination of carbomer or hydroxypropyl methylcellulose (HPMC) as gelling agent and polysorbate 80 as emulsifier. The response of gelling agent type (carbomer or HPMC) and concentration of other two variables penetration enhancer and emulsifier were studied using 2(3)factorial design during in vitro drug release through excised rat skin. Biological activity of emulgel formulation was also investigated using Albino rabbits alone and in combination with iontophoresis. The in vivo efficacy of insulin emulgel was assessed by measuring the blood glucose level at start of the experiment and after every 15 minutes interval for 120 minutes. Total eight formulations were studied. F4 formulation showed maximum insulin permeation flux (4.88 ± 0.09 μg/cm(2)/hour) through excised rat skin. Insulin permeation from these formulations was found to follow the Korsmeyer-Peppas model (r(2) =0.975 to 0.998) during 24 hour with non-Fickian mechanism. Formulation F4 was further investigated in Albino rabbits. For the first group (treated with insulin emulgel alone) the blood glucose level decreased from initial value 250±10mg/dl to 185±7mg/dl at 120 minutes and for the second group (treated with insulin emulgel plus iontophoresis) the blood glucose level decreased to 125±5mg/dl in 120 minutes (P<0.05). It was observed that absorption of insulin through transdermal emulgel was greater in combination with iontophoresis to decrease blood glucose level. On the basis of this study, it has been shown that application of insulin emulgel iontophoretically can be used as alternative (acceptable & painless) to injectable insulin subject to further studies on large animals.
Sigma-Aldrich
Isopropyl myristate, ≥98%
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% methoxyl basis
Sigma-Aldrich
Methyl cellulose, tested according to Ph. Eur.
Sigma-Aldrich
Methyl cellulose, medium viscosity, Methoxyl content 27.5-31.5 %
Sigma-Aldrich
Isopropyl myristate, 98%
Sigma-Aldrich
Methyl cellulose, viscosity: 4,000 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP, BioReagent, suitable for cell culture
Sigma-Aldrich
Methyl cellulose, 26.0-33.0% (Methoxy group (dry basis)), meets USP testing specifications, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, meets USP testing specifications, 26.0-33.0% (methoxyl group, on Dry Basis), viscosity: 400 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 25 cP
Sigma-Aldrich
Methyl cellulose, viscosity: 1,500 cP
Sigma-Aldrich
Methyl cellulose, 27.5-31.5% (Methoxyl content), viscosity: 400 cP
Sigma-Aldrich
Isopropyl myristate, ≥90% (GC)
Sigma-Aldrich
Methyl cellulose, viscosity: 15 cP
Sigma-Aldrich
Methyl cellulose, viscosity 3000-5500 mPa.s, 2 % in H2O(20 °C)
Supelco
Isopropyl myristate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 2,600-5,600 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 40-60 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~90,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, viscosity 80-120 cP, 2 % in H2O(20 °C)(lit.)
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~120,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~10,000
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose, average Mn ~86,000
Sigma-Aldrich
Hypromellose, meets USP testing specifications
Sigma-Aldrich
(Hydroxypropyl)methyl cellulose