MilliporeSigma
  • What's new in lysozyme research? Always a model system, today as yesterday.
What's new in lysozyme research? Always a model system, today as yesterday.
Molecular and cellular biochemistry (1984-09-01)
P Jollès, J Jollès
ABSTRACT
The present review is focused on the main achievements realized in the lysozyme research field since the meeting held in 1972 to commemorate the fiftieth anniversary of the discovery of this enzyme. Despite of extensive structural, physico-chemical, crystallographic, genetic, immunological and evolutionary studies devoted to lysozymes, their biological role is still not exactly known.
Sigma-Aldrich
Lysozyme from chicken egg white, dialyzed, lyophilized, powder, ~100000 U/mg