MilliporeSigma
  • [Bio-based pharmaceutical polymers, possibility of their chemical modification and the applicability of modified polymers].
[Bio-based pharmaceutical polymers, possibility of their chemical modification and the applicability of modified polymers].
Acta pharmaceutica Hungarica (2013-03-01)
István Sebe, Barnabás Szabó, Romána Zelkó
ABSTRACT
Different types of polymers are widely used in biomedical, pharmaceutical and cosmetic purposes. Their applications are curbed, if the polymers can not break down by the body or if the polymer itself is harmful or decompose to harmful material. Authors provide an overview of different types of pharmaceutical polymers of various sources, of the structural characterization and possibilities of their chemical modification and of the classical and instrumental analytical examination methods. The paper deals with the limitations of the use of biopolymers, as well.
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥95% (HPLC)
Millipore
Agar, high purity, low ionic content, low gel strenght, suitable for microbiology
Millipore
Agar, powder, suitable for microbiology
Millipore
Agar, suitable for microbiology
Millipore
Agar, suitable for microbiology
Sigma-Aldrich
N-Acetyl-D-glucosamine, suitable for cell culture, BioReagent
Sigma-Aldrich
N-Acetyl-D-glucosamine, ≥99% (HPLC)
Sigma-Aldrich
Guar
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Sigma-Aldrich
Select agar
Sigma-Aldrich
Agar, Type A, suitable for plant cell culture
Sigma-Aldrich
Agar, microbiology tested, suitable for plant cell culture, suitable for cell culture, powder
Sigma-Aldrich
Agar, Bacteriological, microbiology tested, suitable for plant cell culture, granular
Sigma-Aldrich
Agar, ash 2.0-4.5%
Sigma-Aldrich
Noble agar
Sigma-Aldrich
Agar, High gel strength, suitable for plant cell culture
Sigma-Aldrich
Agar, Type M, suitable for plant cell culture
Sigma-Aldrich
Agar, suitable for plant cell culture, powder
Sigma-Aldrich
Agar, suitable for plant cell culture
Sigma-Aldrich
Polylactic acid, Mw ~60,000
Sigma-Aldrich
Bacteriological agar, for molecular biology
Supelco
Pectin, for use with Total Dietary Fiber Control Kit, TDF-C10