MilliporeSigma
  • Formation of styrene dependent on fermentation management during wheat beer production.
Formation of styrene dependent on fermentation management during wheat beer production.
Food chemistry (2013-02-28)
Katrin J Schwarz, René Stübner, Frank-Jürgen Methner
ABSTRACT
Styrene is formed by the thermal decarboxylation of cinnamic acid during wort boiling or by enzymatic decarboxylation during fermentation. The enzymatic reactions proceed in parallel to the decarboxylation of ferulic- and p-cumaric acid to 4-vinylguaiacol and 4-vinylphenol by the same decarboxylase enzyme. However, the formation of styrene occurs much faster and all available cinnamic acid in wort was converted completely within a few hours. Moreover, the comparison of various manufacturing parameters shows that a higher fermentation temperature of 25 °C compared to 16 °C and an open fermentation management lead to a rapid decrease of styrene. This allows minimising the content of styrene in beer while maintaining the typical wheat beer flavours.
Product Description
Supelco
Styrene, analytical standard
Supelco
Styrene, analytical standard
Sigma-Aldrich
Styrene, ReagentPlus®, contains 4-tert-butylcatechol as stabilizer, ≥99%
Supelco
trans-Cinnamic acid, analytical standard
Sigma-Aldrich
trans-Cinnamic acid, ≥99%, FG
Sigma-Aldrich
trans-Cinnamic acid, natural, ≥99%, FCC, FG
Sigma-Aldrich
trans-Cinnamic acid, ≥99%
Sigma-Aldrich
trans-Cinnamic acid, 97%