MilliporeSigma
Modeling of mixed-mode chromatography of peptides.
Journal of chromatography. A (2013-02-26)
Susanna Bernardi, David Gétaz, Nicola Forrer, Massimo Morbidelli
ABSTRACT
Mixed-mode chromatographic materials are more and more often used for the purification of biomolecules, such as peptides and proteins. In many instances they in fact exhibit better selectivity values and therefore improve the purification efficiency compared to classical materials. In this work, a model to describe biomolecules retention in cation-exchange/reversed-phase (CIEX-RP) mixed-mode columns under diluted conditions has been developed. The model accounts for the effect of the salt and organic modifier concentration on the biomolecule Henry coefficient through three parameters: α, β and γ. The α parameter is related to the adsorption strength and ligand density, β represents the number of organic modifier molecules necessary to displace one adsorbed biomolecule and γ represents the number of salt molecules necessary to desorb one biomolecule. The latter parameter is strictly related to the number of charges on the biomolecule surface interacting with the ion-exchange ligands and it is shown experimentally that its value is close to the biomolecule net charge. The model reliability has been validated by a large set of experimental data including retention times of two different peptides (goserelin and insulin) on five columns: a reversed-phase C8 column and four CIEX-RP columns with different percentages of sulfonic groups and various concentration values of the salt and organic modifier. It has been found that the percentage of sulfonic groups on the surface strongly affects the peptides adsorption strength, and in particular, in the cases investigated, a CIEX ligand density around 0.04μmol/m(2) leads to optimal retention values.
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (w/v) ammonium formate, 5 % (v/v) water, 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium acetate solution, BioUltra, for molecular biology, ~3 M in H2O
Sigma-Aldrich
Sodium acetate, 99.995% trace metals basis
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) trifluoroacetic acid, suitable for HPLC
Sigma-Aldrich
Acetonitrile solution, contains 0.05 % (v/v) trifluoroacetic acid
Sigma-Aldrich
Acetonitrile solution, contains 0.1 % (v/v) formic acid, suitable for HPLC
Sigma-Aldrich
Sodium acetate, anhydrous, BioUltra, for luminescence, for molecular biology, ≥99.0% (NT)
Sigma-Aldrich
Sodium acetate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium acetate, anhydrous, for molecular biology, ≥99%
Sigma-Aldrich
Sodium acetate, powder, BioReagent, suitable for electrophoresis, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium acetate, meets USP testing specifications, anhydrous
Sigma-Aldrich
Acetonitrile solution, contains 10.0% acetone, 0.05% formic acid, 40.0% 2-propanol
Sigma-Aldrich
Sodium acetate, anhydrous, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate, puriss. p.a., ACS reagent, reag. Ph. Eur., anhydrous
Sigma-Aldrich
Sodium acetate, ACS reagent, ≥99.0%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Sodium acetate trihydrate, ReagentPlus®, ≥99.0%
Sigma-Aldrich
Sodium acetate trihydrate, puriss., meets analytical specification of Ph. Eur., BP, USP, FCC, E262, 99.0-101.0% (calc. to the dried substance), ≤0.00002% Al
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.5% (GC)
Sigma-Aldrich
Sodium acetate trihydrate, ACS reagent, ≥99%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%, poly-coated bottles
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ≥99.5%, ACS reagent
Supelco
Acetonitrile, analytical standard
Supelco
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9% (GC)
Sigma-Aldrich
Sodium acetate trihydrate, BioXtra, ≥99.0%