MilliporeSigma
  • A reversible fluorescent probe for detecting hypochloric acid in living cells and animals: utilizing a novel strategy for effectively modulating the fluorescence of selenide and selenoxide.
A reversible fluorescent probe for detecting hypochloric acid in living cells and animals: utilizing a novel strategy for effectively modulating the fluorescence of selenide and selenoxide.
Chemical communications (Cambridge, England) (2013-02-19)
Zhangrong Lou, Peng Li, Qiang Pan, Keli Han
ABSTRACT
Based on a novel strategy for modulating the fluorescence of selenide and selenoxide, we have designed and developed a reversible fluorescent probe for hypochloric acid. And the synthesis, characterization, fluorescence properties, as well as the biological applications in living cells and animals, have all been described.
Sigma-Aldrich
Selenium dioxide, ReagentPlus®, powder, 99.8% trace metals basis
Sigma-Aldrich
Selenium dioxide, 99.999% trace metals basis
Sigma-Aldrich
Selenium dioxide, reagent grade, powder, 98%
Sigma-Aldrich
Selenium dioxide, ≥99.9% trace metals basis