MilliporeSigma
  • Synthesis and characterization of a molecularly imprinted polymer for the determination of trace tributyltin in seawater and seafood by liquid chromatography-tandem mass spectroscopy.
Synthesis and characterization of a molecularly imprinted polymer for the determination of trace tributyltin in seawater and seafood by liquid chromatography-tandem mass spectroscopy.
Journal of chromatography. B, Analytical technologies in the biomedical and life sciences (2013-02-19)
Shanshan Zhu, Futao Hu, Ting Yang, Ning Gan, Daodong Pan, Yuting Cao, Dazhen Wu
ABSTRACT
Analysis of tributyltin chloride (TBT) in environmental samples, such as seawater, is important in order to evaluate the TBT contamination and accumulation in the trophic chain. The environmental impact of organotin compounds has been a particular focus of analytical studies. The present study reports the use of molecular imprinting technology coupled with liquid chromatography-tandem mass spectrometry (LC-MS/MS) to determine trace amounts of TBT in seawater and seafood (mussel tissue samples). The imprinted polymer was synthesized by a non-covalent free-radical approach using acrylamide (AM) as a monomer and TBT as a template molecule in acetonitrile solvent (polymerization media). The imprinted polymer synthesized by this approach exhibited good adsorptive capacity and allowed specific retention of TBT. Recoveries of TBT in seawater samples spiked with different TBT concentrations ranged from 67.2% to 81.1% with peak area precision (RSD)<3.7%, and recoveries of TBT in mussel tissue samples ranged from 75.0% to 94.2% with RSD<4.8%.
Sigma-Aldrich
Tributyltin hydride, contains 0.05% BHT as stabilizer, 97%