MilliporeSigma
  • Fast and continuous preparation of high polymerization degree cellulose nanofibrils and their three-dimensional macroporous scaffold fabrication.
Fast and continuous preparation of high polymerization degree cellulose nanofibrils and their three-dimensional macroporous scaffold fabrication.
Nanoscale (2013-02-16)
Jiankang Song, Aimin Tang, Tingting Liu, Jufang Wang
ABSTRACT
C6-carboxy-cellulose with a carboxylate content of 0.8 mmol g(-1) was obtained by oxidation of once-dried cellulose, using the 2,2,6,6-tetramethylpiperidinyl-1-oxyl (TEMPO)/NaClO/NaClO2 system at pH 6.8 and 60 °C for 16 h. This method, with the addition of reagents in the order TEMPO, NaClO and NaClO2, was 38 h faster than a previously published method. Individualized cellulose nanofibrils with a width of 3-5 nm and a length of several hundred nanometers were prepared by homogenizing the C6-carboxy-cellulose-water suspension. Macroporous cellulose nanofibril/poly(vinyl alcohol) scaffolds with interconnected large pores of 20-100 μm diameter and small pores of 2-10 μm diameter were fabricated. The cellulose nanofilaments formed nanofibrous structures on the surface of the PVA wall, which was similar to that of the collagen skeleton of the extracellular matrix. NIH/3T3 cells were cultured in the scaffolds for 4 weeks, SEM observation showed that the cells were anchored and clustered on the cellulose nanofilaments, forming spherical colonies. The extracellular matrix (ECM) was filled with mineralized particles.
Sigma-Aldrich
Poly(vinyl alcohol), Mw 9,000-10,000, 80% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 13,000-23,000, 98% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 6-98, Mw ~47,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 10-98, Mw ~61,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 13,000-23,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 4-88, Mw ~31,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 4-98, Mw ~27,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 85,000-124,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 146,000-186,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 31,000-50,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 89,000-98,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 146,000-186,000, 87-89% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 85,000-124,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), 87-90% hydrolyzed, average mol wt 30,000-70,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Fully hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 8-88, Mw ~67,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), Mw 31,000-50,000, 98-99% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Poly(vinyl alcohol), average Mw 130,000, 99+% hydrolyzed
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 20-98, Mw ~125,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chlorate, ACS reagent, ≥99.0%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium chlorate, ReagentPlus®, ≥99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Kuraray Poval ®56-98, Mw ~195,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 28-99, Mw ~145,000
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 40-88, average Mw ~205,000 g/mol
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Perchloric acid, 70%, 99.999% trace metals basis
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TEMPO, 98%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
TEMPO, purified by sublimation, 99%
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Mowiol® 18-88, Mw ~130,000
SKU
Pack Size
Availability
Price
Quantity
Supelco
Perchloric acid, 0.01 M HClO4 in water (0.01N), eluent for IC
SKU
Pack Size
Availability
Price
Quantity
Sigma-Aldrich
Sodium perchlorate, ACS reagent, ≥98.0%