MilliporeSigma
  • Peyer's patch-mediated intestinal immune system modulating activity of pectic-type polysaccharide from peel of Citrus unshiu.
Peyer's patch-mediated intestinal immune system modulating activity of pectic-type polysaccharide from peel of Citrus unshiu.
Food chemistry (2013-02-16)
Hyung-Joo Suh, Hyun-Seuk Yang, Kyung-Soo Ra, Dong-Ouk Noh, Ki-Han Kwon, Jong-Hyun Hwang, Kwang-Won Yu
ABSTRACT
An intestinal immune system modulating polysaccharide (CUI-3IIb-3-2, 18kDa) was purified from Citrus unshiu peel. CUI-3IIb-3-2 mainly comprised GalA, GlcA, Ara, Gal and Rha, and it consisted of 4-linked GalA, terminal Araf, 4- or 5-linked/3,4- or 3,5-branched Ara, terminal Gal, and 2-linked/2,4-branched Rha. After CUI-3IIb-3-2 digestion by endo-α-d-(1→4)-polygalacturonase, its hydrolysate was fractionated into PG-1 and PG-2. Methylation analyses of PG-1 and PG-2 using base-catalysed β-elimination suggested that CUI-3IIb-3-2 be assumed as pectic-type polysaccharide. Since the activities of PG-1 and PG-2 were potently decreased, the whole polysaccharide structure of CUI-3IIb-3-2 would be essential to maintain the activity. Meanwhile, when CUI-3IIb was orally administered in mice, bone marrow cell proliferation and GM-CSF/IL-6 production from Peyer's patch cell were significantly higher (1.76- and 2.03/2.51-fold, respectively) than a saline. Therefore, a pectic-type polysaccharide from citrus peel could stimulate Peyer's patches and produce hematopoietic growth factors resulted in bone marrow cell proliferation.
Sigma-Aldrich
Pectin from citrus peel, Galacturonic acid ≥74.0 % (dried basis)
Supelco
Pectin, for use with Total Dietary Fiber Control Kit, TDF-C10