MilliporeSigma
  • [Relationship of gastric microflora and neopterin content in gastric juice in children with chronic gastroduodenitis].
[Relationship of gastric microflora and neopterin content in gastric juice in children with chronic gastroduodenitis].
Eksperimental'naia i klinicheskaia gastroenterologiia = Experimental & clinical gastroenterology (2013-02-14)
N Iu Kon'kova
ABSTRACT
The study included 71 children with chronic gastroduodenitis aged 5-17 years. PCR DNA was determined by the presence of Helicobacter pylori, human papilloma virus high cancer risk 16, 18 types (HPV), herpes simplex 1 and type 2 (HSV), cytomegalovirus (CMV), fungi, Candida in gastric juice and biopsy specimens of gastric mucosa and duodenal. Viruses were detected in 14% of patients, an association of microorganisms - in 20% of the children, the isolated H. pylori infection - 18%. The relationship between the composition of microflora in the gastroduodenal region and neopterin levels in gastric juice with a distinct increase in its value in the presence of viruses.
Sigma-Aldrich
D-(+)-Neopterin, ≥97.5% (sum of enantiomers, HPLC)