MilliporeSigma
  • Fast and interference-free determination of glyphosate and glufosinate residues through electrophoresis in disposable microfluidic chips.
Fast and interference-free determination of glyphosate and glufosinate residues through electrophoresis in disposable microfluidic chips.
Journal of chromatography. A (2013-02-13)
Xuan Wei, Xiaotong Gao, Lei Zhao, Xianglu Peng, Lei Zhou, Jing Wang, Qiaosheng Pu
ABSTRACT
With an increasing concern on food safety, fast screening of residues of widespread herbicides becomes necessary. Herein we report a microchip electrophoresis system with laser induced fluorescence (LIF) detection for rapid and sensitive analysis of glyphosate (GLYP) and glufosinate (GLUF) residues. Disposable cyclic olefin copolymer microchips and a low-cost LIF detector were employed to minimize the cost of the analysis. Systematic optimization of experimental conditions was performed to achieve highly efficient analysis. Under the selected condition, GLYP and GLUF were efficiently resolved from sample matrices with a buffer containing 10 mmol/L borax and 2.0% (m/v) hydroxypropyl cellulose at pH 9.0. The number of theoretical plates of 1.0×10(6) m(-1) was attained for both analytes. Derivatization at lower concentrations (<10 μg/L) was also examined, successful detection of 0.34 μg/L GLYP and 0.18 μg/L GLUF was confirmed. The system was applied for the determination of both analytes in real samples without any preconcentration involved. Recoveries of GLYP and GLUF spiked in these samples were 84.0-101.0% and 90.0-103.0%, respectively.
Sigma-Aldrich
N-(Phosphonomethyl)glycine, 96%
Sigma-Aldrich
N-(Phosphonomethyl)glycine, BioReagent, suitable for plant cell culture
Supelco
Glyphosate, PESTANAL®, analytical standard
Sigma-Aldrich
Sodium tetraborate decahydrate, BioXtra, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, puriss., meets analytical specification of Ph. Eur., BP, NF, 99.0-103.0%
Sigma-Aldrich
Sodium tetraborate decahydrate, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, ACS reagent, ≥99.5%
Sigma-Aldrich
Sodium tetraborate decahydrate, puriss. p.a., ACS reagent, reag. ISO, buffer substance, ≥99.5%